2-Brom-5-nitrobenzonitril
Lieferant: COMBI-BLOCKS
Synonyme:
1-Cyano-2-bromo-5-nitrobenzene
AN-1149-5G
AN-1149-25G
AN-1149-1G
COBBAN-1149-5GEA
96.3
EUR
COBBAN-1149-5G
COBBAN-1149-25G
COBBAN-1149-1G
2-Brom-5-nitrobenzonitril
2-Brom-5-nitrobenzonitril
Formel:
C₇H₃BrN₂O₂ Molecular Weight: 227.02 g/mol |
MDL:
MFCD00234249 CAS-Nummer: 134604-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...