Methyl-3-brom-4-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromo-4-fluorobenzoate

CA-4099-5G CA-4099-25G CA-4099-1G
COBBCA-4099-5GEA 101 EUR
COBBCA-4099-5G COBBCA-4099-25G COBBCA-4099-1G
Methyl-3-brom-4-fluorbenzoat
Methyl-3-brom-4-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233.04 g/mol
MDL: MFCD04972915
CAS-Nummer: 82702-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR