Methyl-4-brom-3-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-bromo-3-fluorobenzoate

CA-4107-25G CA-4107-5G CA-4107-1G
COBBCA-4107-25GEA 514 EUR
COBBCA-4107-25G COBBCA-4107-5G COBBCA-4107-1G
Methyl-4-brom-3-fluorbenzoat
Methyl-4-brom-3-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233.04 g/mol
MDL: MFCD08436044
CAS-Nummer: 849758-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR