4-Brom-3-hydroxybenzoesäure

Lieferant: COMBI-BLOCKS

CA-4188-5G CA-4188-1G
COBBCA-4188-5GEA 53.4 EUR
COBBCA-4188-5G COBBCA-4188-1G
4-Brom-3-hydroxybenzoesäure
4-Bromo-3-hydroxybenzoic acid
Formel: C₇H₅BrO₃
MDL: MFCD08056371
CAS-Nummer: 14348-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR