Methyl-2-brom-4-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-4-fluorobenzoate

CA-4306-1G CA-4306-25G CA-4306-5G
COBBCA-4306-1GEA 25.9 EUR
COBBCA-4306-1G COBBCA-4306-25G COBBCA-4306-5G
Methyl-2-brom-4-fluorbenzoat
Methyl-2-brom-4-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233,04 g/mol
MDL: MFCD06204026
CAS-Nummer: 653-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR