Methyl-2-brom-5-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-5-fluorobenzoate

CA-4310-5G CA-4310-25G CA-4310-1G CA-4310-100G
COBBCA-4310-5GEA 43.4 EUR
COBBCA-4310-5G COBBCA-4310-25G COBBCA-4310-1G COBBCA-4310-100G
Methyl-2-brom-5-fluorbenzoat
Methyl-2-brom-5-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233.04 g/mol
MDL: MFCD00086167
CAS-Nummer: 6942-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR