3-Formyl-6-methoxypyridin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde
CA-4314-5G
CA-4314-1G
CA-4314-25G
COBBCA-4314-5GEA
70.8
EUR
COBBCA-4314-5G
COBBCA-4314-1G
COBBCA-4314-25G
3-Formyl-6-methoxypyridin
3-Formyl-6-methoxypyridin
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol |
MDL:
MFCD02683446 CAS-Nummer: 65873-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...