5-(Boc-amino)-2-brompyridin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (6-bromopyridin-3-yl)carbamate, 5-Amino-2-bromopyridine, 5-BOC protected

CA-4319-5G CA-4319-1G
COBBCA-4319-5GEA 387 EUR
COBBCA-4319-5G COBBCA-4319-1G
5-(Boc-amino)-2-brompyridin
5-(Boc-amino)-2-brompyridin
Formel: C₁₀H₁₃BrN₂O₂
Molecular Weight: 273,13 g/mol
MDL: MFCD07363808
CAS-Nummer: 218594-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR