Methyl-3-fluor-5-hydroxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-fluoro-5-hydroxybenzoate

CA-4339-5G CA-4339-1G CA-4339-250M
COBBCA-4339-5GEA 1880 EUR
COBBCA-4339-5G COBBCA-4339-1G COBBCA-4339-250M
Methyl-3-fluor-5-hydroxybenzoat
Methyl-3-fluor-5-hydroxybenzoat
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
MDL: MFCD14698043
CAS-Nummer: 1072004-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR