3-Brom-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

CA-4409-5G CA-4409-1G
COBBCA-4409-5GEA 812 EUR
COBBCA-4409-5G COBBCA-4409-1G
3-Brom-5-nitrobenzonitril
3-Brom-5-nitrobenzonitril
Formel: C₇H₃BrN₂O₂
Molecular Weight: 227.02 g/mol
MDL: MFCD00463804
CAS-Nummer: 49674-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR