Ethyl-2-brom-4-chlorbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-bromo-4-chlorobenzoate
CA-4491-5G
CA-4491-1G
COBBCA-4491-5GEA
356
EUR
COBBCA-4491-5G
COBBCA-4491-1G
Ethyl-2-brom-4-chlorbenzoat
Ethyl-2-brom-4-chlorbenzoat
Formel:
C₉H₈BrClO₂ Molecular Weight: 263,52 g/mol |
MDL:
MFCD10566824 CAS-Nummer: 690260-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...