Ethyl-3-brom-4-fluorbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 3-bromo-4-fluorobenzoate
CA-4525-5G
CA-4525-1G
CA-4525-25G
COBBCA-4525-5GEA
114
EUR
COBBCA-4525-5G
COBBCA-4525-1G
COBBCA-4525-25G
Ethyl-3-brom-4-fluorbenzoat
Ethyl-3-brom-4-fluorbenzoat
Formel:
C₉H₈BrFO₂ Molecular Weight: 247,06 g/mol |
MDL:
MFCD06204378 CAS-Nummer: 23233-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...