Methyl-3-brom-5-hydroxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromo-5-hydroxybenzoate

CA-4579-5G CA-4579-1G
COBBCA-4579-5GEA 946 EUR
COBBCA-4579-5G COBBCA-4579-1G
Methyl-3-brom-5-hydroxybenzoat
Methyl-3-brom-5-hydroxybenzoat
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD10566800
CAS-Nummer: 192810-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR