Methyl-2-(trifluormethyl)benzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-(trifluoromethyl)benzoate
CA-4626-5G
CA-4626-25G
CA-4626-1G
COBBCA-4626-5GEA
48.4
EUR
COBBCA-4626-5G
COBBCA-4626-25G
COBBCA-4626-1G
Methyl-2-(trifluormethyl)benzoat
Methyl-2-(trifluormethyl)benzoat
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol |
MDL:
MFCD00236050 CAS-Nummer: 344-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...