Methyl-3-(trifluormethyl)benzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 3-(trifluoromethyl)benzoate
CA-4627-5G
CA-4627-25G
COBBCA-4627-5GEA
0
EUR
COBBCA-4627-5G
COBBCA-4627-25G
Methyl-3-(trifluormethyl)benzoat
Methyl-3-(trifluormethyl)benzoat
Formel:
C₉H₇F₃O₂ |
MDL:
MFCD00043543 CAS-Nummer: 2557-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...