Methyl-4-(trifluormethyl)benzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-(trifluoromethyl)benzoate

CA-4628-5G CA-4628-25G CA-4628-100G
COBBCA-4628-5GEA 0 EUR
COBBCA-4628-5G COBBCA-4628-25G COBBCA-4628-100G
Methyl-4-(trifluormethyl)benzoat
Methyl-4-(trifluormethyl)benzoat
Formel: C₉H₇F₃O₂
MDL: MFCD00042324
CAS-Nummer: 2967-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR