Methyl-3-(trifluormethoxy)benzoat

Lieferant: COMBI-BLOCKS

CA-4657-5G CA-4657-25G CA-4657-1G
COBBCA-4657-5GEA 115 EUR
COBBCA-4657-5G COBBCA-4657-25G COBBCA-4657-1G
Methyl-3-(trifluormethoxy)benzoat
Methyl-3-(trifluormethoxy)benzoat
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
MDL: MFCD00041005
CAS-Nummer: 148438-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR