Methyl-2,3-dichlorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2,3-dichlorobenzoate, 2,3-Dichlorobenzoic acid methyl ester

CA-4671-5G CA-4671-1G CA-4671-25G
COBBCA-4671-5GEA 56 EUR
COBBCA-4671-5G COBBCA-4671-1G COBBCA-4671-25G
Methyl-2,3-dichlorbenzoat
Methyl-2,3-dichlorbenzoat
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00061099
CAS-Nummer: 2905-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR