Methyl-2-brom-5-(trifluormethoxy)benzoat

Lieferant: COMBI-BLOCKS

CA-4674-1G CA-4674-5G
COBBCA-4674-1GEA 303 EUR
COBBCA-4674-1G COBBCA-4674-5G
Methyl-2-brom-5-(trifluormethoxy)benzoat
Methyl-2-brom-5-(trifluormethoxy)benzoat
Formel: C₉H₆BrF₃O₃
Molecular Weight: 299,04 g/mol
MDL: MFCD12025999
CAS-Nummer: 1150114-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR