Methyl-4-(trifluormethoxy)benzoat

Lieferant: COMBI-BLOCKS

CA-4708-1G CA-4708-5G CA-4708-25G
COBBCA-4708-1GEA 25.5 EUR
COBBCA-4708-1G COBBCA-4708-5G COBBCA-4708-25G
Methyl-4-(trifluormethoxy)benzoat
Methyl-4-(trifluormethoxy)benzoat
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
MDL: MFCD00041006
CAS-Nummer: 780-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR