Ethyl-5-brom-2,3-difluorbenzoat

Lieferant: COMBI-BLOCKS

CA-4731-5G CA-4731-1G
COBBCA-4731-5GEA 1590 EUR
COBBCA-4731-5G COBBCA-4731-1G
Ethyl-5-brom-2,3-difluorbenzoat
Ethyl-5-brom-2,3-difluorbenzoat
Formel: C₉H₇BrF₂O₂
MDL: MFCD12756425
CAS-Nummer: 1187386-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR