Methyl-2-brom-6-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-6-fluorobenzoate, 2-Bromo-6-fluorobenzoic acid methyl ester

CA-4964-5G CA-4964-1G
COBBCA-4964-5GEA 83.4 EUR
COBBCA-4964-5G COBBCA-4964-1G
Methyl-2-brom-6-fluorbenzoat
Methyl-2-brom-6-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233.04 g/mol
MDL: MFCD10000922
CAS-Nummer: 820236-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR