Methyl-3-brom-4-hydroxy-5-methoxybenzoat

Lieferant: COMBI-BLOCKS

CA-4974-25G CA-4974-5G CA-4974-1G
COBBCA-4974-25GEA 1180 EUR
COBBCA-4974-25G COBBCA-4974-5G COBBCA-4974-1G
Methyl-3-brom-4-hydroxy-5-methoxybenzoat
Methyl-3-brom-4-hydroxy-5-methoxybenzoat
Formel: C₉H₉BrO₄
Molecular Weight: 261,07 g/mol
MDL: MFCD06203663
CAS-Nummer: 108249-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR