3-Brom-5-butoxybenzoesäure

Lieferant: COMBI-BLOCKS

CA-4993-5G CA-4993-1G
COBBCA-4993-5GEA 595 EUR
COBBCA-4993-5G COBBCA-4993-1G
3-Brom-5-butoxybenzoesäure
3-Bromo-5-butoxybenzoic acid
Formel: C₁₁H₁₃BrO₃
MDL: MFCD16618945
CAS-Nummer: 1228956-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR