2-Brom-6-ethoxypyridin

Lieferant: COMBI-BLOCKS

HC-6405-1G HC-6405-25G HC-6405-5G
COBBHC-6405-1GEA 137 EUR
COBBHC-6405-1G COBBHC-6405-25G COBBHC-6405-5G
2-Brom-6-ethoxypyridin
2-Bromo-6-ethoxypyridine
Formel: C₇H₈BrNO
Molecular Weight: 202.05 g/mol
MDL: MFCD00234958
CAS-Nummer: 4645-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR