2-Brom-6-propoxypyridin

Lieferant: COMBI-BLOCKS

HC-6451-5G HC-6451-1G HC-6451-25G
COBBHC-6451-5GEA 312 EUR
COBBHC-6451-5G COBBHC-6451-1G COBBHC-6451-25G
2-Brom-6-propoxypyridin
2-Bromo-6-propoxypyridine
Formel: C₈H₁₀BrNO
MDL: MFCD00234429
CAS-Nummer: 891842-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR