2-Brom-6-isopropoxypyridin

Lieferant: COMBI-BLOCKS

HC-6487-1G HC-6487-25G HC-6487-5G
COBBHC-6487-1GEA 204 EUR
COBBHC-6487-1G COBBHC-6487-25G COBBHC-6487-5G
2-Brom-6-isopropoxypyridin
2-Brom-6-isopropoxypyridin
Formel: C₈H₁₀BrNO
MDL: MFCD09878949
CAS-Nummer: 463336-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR