Methyl-3-furoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl furan-3-carboxylate

HI-1506-5G HI-1506-25G HI-1506-1G
COBBHI-1506-5GEA 114 EUR
COBBHI-1506-5G COBBHI-1506-25G COBBHI-1506-1G
Methyl-3-furoat
Methyl-3-furoat
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
MDL: MFCD06203671
CAS-Nummer: 13129-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR