4-Chlorpyridin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloropyridinium chloride

OS-7402-25G OS-7402-100G
COBBOS-7402-25GEA 38.2 EUR
COBBOS-7402-25G COBBOS-7402-100G
4-Chlorpyridin Hydrochlorid
4-Chlorpyridin Hydrochlorid
Formel: C₅H₅Cl₂N
MDL: MFCD00012829
CAS-Nummer: 7379-35-3
EINECS: 230-946-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR