2-Picolylchlorid Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Chloromethyl)pyridine HCl, 2-(Chloromethyl)pyridine hydrochloride

OS-7431-100G OS-7431-25G
COBBOS-7431-100GEA 127 EUR
COBBOS-7431-100G COBBOS-7431-25G
2-Picolylchlorid Hydrochlorid
2-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
Molecular Weight: 164.03 g/mol
MDL: MFCD00012811
CAS-Nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR