3-Furancarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Furan-3-carboxylic acid, 3-Furancarboxylic acid
OS-7504-25G
OS-7504-5G
OS-7504-100G
COBBOS-7504-25GEA
153
EUR
COBBOS-7504-25G
COBBOS-7504-5G
COBBOS-7504-100G
3-Furancarbonsäure
3-Furoic acid
Formel:
C₅H₄O₃ Molecular Weight: 112,08 g/mol |
MDL:
MFCD00005350 CAS-Nummer: 488-93-7 EINECS: 207-689-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...