Bis(triphenylphosphin)palladium(II)chlorid

Lieferant: COMBI-BLOCKS

Synonyme: Bis(triphenylphosphine)palladium(II) dichloride

OS-7517-5G OS-7517-25G OS-7517-1G OS-7517-100G
COBBOS-7517-5GEA 471 EUR
COBBOS-7517-5G COBBOS-7517-25G COBBOS-7517-1G COBBOS-7517-100G
Bis(triphenylphosphin)palladium(II)chlorid
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Molecular Weight: 701,91 g/mol
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR