2-Chlor-3-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

OS-7694-5G OS-7694-25G OS-7694-1G
COBBOS-7694-5GEA 50.8 EUR
COBBOS-7694-5G COBBOS-7694-25G COBBOS-7694-1G
2-Chlor-3-formylpyridin
2-Chlor-3-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141,56 g/mol
MDL: MFCD01315308
CAS-Nummer: 36404-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR