2-(Boc-amino)-4-chlorpyridin

Lieferant: COMBI-BLOCKS

OT-0177-1G OT-0177-5G OT-0177-25G
COBBOT-0177-1GEA 0 EUR
COBBOT-0177-1G COBBOT-0177-5G COBBOT-0177-25G
2-(Boc-amino)-4-chlorpyridin
2-(Boc-amino)-4-chlorpyridin
Formel: C₁₀H₁₃ClN₂O₂
MDL: MFCD05663519
CAS-Nummer: 130721-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR