2-Acetamido-6-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: N-(6-Methylpyridin-2-yl)acetamide

OT-0180-5G OT-0180-1G
COBBOT-0180-5GEA 63.9 EUR
COBBOT-0180-5G COBBOT-0180-1G
2-Acetamido-6-methylpyridin
2-Acetamido-6-methylpyridin
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
MDL: MFCD00091881
CAS-Nummer: 5327-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR