3-Brom-4-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

OT-8106-1G OT-8106-10G
COBBOT-8106-1GEA 51.3 EUR
COBBOT-8106-1G COBBOT-8106-10G
3-Brom-4-formylpyridin
3-Brom-4-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD05864506
CAS-Nummer: 70201-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR