3-Brom-2-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromopyridine-2-carboxaldehyde

OT-8108-5G OT-8108-1G
COBBOT-8108-5GEA 748 EUR
COBBOT-8108-5G COBBOT-8108-1G
3-Brom-2-formylpyridin
3-Brom-2-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD07781220
CAS-Nummer: 405174-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR