2-Chlor-4-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde

OT-8130-5G OT-8130-1G
COBBOT-8130-5GEA 114 EUR
COBBOT-8130-5G COBBOT-8130-1G
2-Chlor-4-formylpyridin
2-Chlor-4-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141,56 g/mol
MDL: MFCD06651557
CAS-Nummer: 101066-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR