3-Chlor-5-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloro-pyridine-3-carbaldehyde, 5-Chloronicotinaldehyde

OT-8131-1G OT-8131-5G
COBBOT-8131-1GEA 141 EUR
COBBOT-8131-1G COBBOT-8131-5G
3-Chlor-5-formylpyridin
3-Chlor-5-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141,56 g/mol
MDL: MFCD08457773
CAS-Nummer: 113118-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR