3-Chlor-2-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloropyridine-2-carboxaldehyde

OT-8132-5G OT-8132-1G
COBBOT-8132-5GEA 408 EUR
COBBOT-8132-5G COBBOT-8132-1G
3-Chlor-2-formylpyridin
3-Chlor-2-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141.56 g/mol
MDL: MFCD09991855
CAS-Nummer: 206181-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR