3-Fluor-5-formylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Fluoronicotinaldehyde
OT-8137-5G
OT-8137-1G
COBBOT-8137-5GEA
262
EUR
COBBOT-8137-5G
COBBOT-8137-1G
3-Fluor-5-formylpyridin
3-Fluor-5-formylpyridin
Formel:
C₆H₄FNO Molecular Weight: 125,1 g/mol |
MDL:
MFCD07778429 CAS-Nummer: 39891-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...