3-Fluor-2-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoropyridine-2-carboxaldehyde, 3-Fluoropicolinaldehyde, 3-Fluoropyridine-2-carbaldehyde, 3-Fluoro-2-pyridinecarboxaldehyde

OT-8138-1G OT-8138-5G
COBBOT-8138-1GEA 47.5 EUR
COBBOT-8138-1G COBBOT-8138-5G
3-Fluor-2-formylpyridin
3-Fluor-2-formylpyridin
Formel: C₆H₄FNO
Molecular Weight: 125,1 g/mol
MDL: MFCD07781234
CAS-Nummer: 31224-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR