5-Fluor-2-formylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Fluoropyridine-2-carboxaldehyde, 5-Fluoropicolinaldehyde
OT-8139-1G
OT-8139-5G
COBBOT-8139-1GEA
181
EUR
COBBOT-8139-1G
COBBOT-8139-5G
5-Fluor-2-formylpyridin
5-Fluor-2-formylpyridin
Formel:
C₆H₄FNO Molecular Weight: 125,1 g/mol |
MDL:
MFCD07781242 CAS-Nummer: 31181-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...