2-Fluor-3-formylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoronicotinaldehyde

PY-1260-1G PY-1260-25G
COBBPY-1260-1GEA 31 EUR
COBBPY-1260-1G COBBPY-1260-25G
2-Fluor-3-formylpyridin
2-Fluor-3-formylpyridin
Formel: C₆H₄FNO
Molecular Weight: 125.1 g/mol
MDL: MFCD03095270
CAS-Nummer: 36404-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR