2-(Boc-amino)-4-brompyridin
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl (4-bromopyridin-2-yl)carbamate
PY-1372-5G
PY-1372-1G
COBBPY-1372-5GEA
334
EUR
COBBPY-1372-5G
COBBPY-1372-1G
2-(Boc-amino)-4-brompyridin
2-(Boc-amino)-4-brompyridin
Formel:
C₁₀H₁₃BrN₂O₂ Molecular Weight: 273.13 g/mol |
MDL:
MFCD09037866 CAS-Nummer: 207799-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...