2-Amino-3,5-dibrom-6-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dibromo-6-methyl-2-pyridinamine

PY-1390-5G PY-1390-25G
COBBPY-1390-5GEA 26.2 EUR
COBBPY-1390-5G COBBPY-1390-25G
2-Amino-3,5-dibrom-6-methylpyridin
2-Amino-3,5-dibromo-6-methylpyridine
Formel: C₆H₆Br₂N₂
Molecular Weight: 265,94 g/mol
MDL: MFCD00068229
CAS-Nummer: 91872-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR