3-Brom-5-formylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde
PY-7244-1G
PY-7244-25G
PY-7244-5G
COBBPY-7244-1GEA
25.9
EUR
COBBPY-7244-1G
COBBPY-7244-25G
COBBPY-7244-5G
3-Brom-5-formylpyridin
3-Brom-5-formylpyridin
Formel:
C₆H₄BrNO Molecular Weight: 186,01 g/mol |
MDL:
MFCD03265758 CAS-Nummer: 113118-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...