2-Amino-3,5-dichlor-6-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Amino-3,5-dichloro-2-methylpyridine

PY-7262-1G PY-7262-25G PY-7262-5G
COBBPY-7262-1GEA 25.5 EUR
COBBPY-7262-1G COBBPY-7262-25G COBBPY-7262-5G
2-Amino-3,5-dichlor-6-methylpyridin
2-Amino-3,5-dichloro-6-methylpyridine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,03 g/mol
MDL: MFCD00129029
CAS-Nummer: 22137-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR