5-Chlor-3-fluor-2-methoxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-chloro-3-fluoropyridine

PY-7298-1G PY-7298-5G
COBBPY-7298-1GEA 86.2 EUR
COBBPY-7298-1G COBBPY-7298-5G
5-Chlor-3-fluor-2-methoxypyridin
5-Chlor-3-fluor-2-methoxypyridin
Formel: C₆H₅ClFNO
Molecular Weight: 161,56 g/mol
MDL: MFCD07375056
CAS-Nummer: 886374-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR