2-Chlor-3-methoxy-6-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-3-methoxy-6-nitropyridine

PY-7323-1G PY-7323-5G
COBBPY-7323-1GEA 208 EUR
COBBPY-7323-1G COBBPY-7323-5G
2-Chlor-3-methoxy-6-nitropyridin
2-Chlor-3-methoxy-6-nitropyridin
Formel: C₆H₅BrN₂O₃
Molecular Weight: 233.02 g/mol
MDL: MFCD06662266
CAS-Nummer: 76066-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR